missing translation for 'onlineSavingsMsg'
Learn More

Ferric Chloride, 41 DEG Be Solution, Spectrum™ Chemical

Catalog No. p-7165545
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
55 gal.
Packaging:
Poly Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610489 500 mL Poly Drum
18610490 4 L Poly Drum
18610487 20 L Poly Drum
18610488 55 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18610489 Supplier Spectrum Chemical Mfg Cor Supplier No. F1025500MLPL
Only null left

Chemical Identifiers

CAS 7705-08-0 , 7732-18-5

Specifications

Color Yellow Brown
CAS Max % 40, 60
Grade Reagent
Packaging Poly Drum
Quantity 500 mL
Chemical Name or Material Ferric Chloride

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.