CAS RN 7705-08-0
Ferric Chloride Solution, 0.025% w/v (APHA), Fisher Chemical
CAS: 7705-08-0 Molecular Formula: Cl3Fe Molecular Weight (g/mol): 162.20 MDL Number: MFCD00011005 InChI Key: RBTARNINKXHZNM-UHFFFAOYSA-K Synonym: Iron (III) Chloride Solution, 0.025% w/v IUPAC Name: iron(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Fe+3]
Iron(III) chloride, 98%, pure, anhydrous
CAS: 7705-08-0 Molecular Formula: Cl3Fe Molecular Weight (g/mol): 162.20 MDL Number: MFCD00011005 InChI Key: RBTARNINKXHZNM-UHFFFAOYSA-K Synonym: Ferric chloride IUPAC Name: iron(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Fe+3]
Iron(III) chloride, anhydrous, 98%
CAS: 7705-08-0 Molecular Formula: Cl3Fe Molecular Weight (g/mol): 162.20 MDL Number: MFCD00011005 InChI Key: RBTARNINKXHZNM-UHFFFAOYSA-K Synonym: Ferric chloride IUPAC Name: iron(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Fe+3]
VeriSpec™ Iron Standard for ICP 10 g/L in 5% HCl, Ricca Chemical
CAS: 7705-08-0 Molecular Formula: Cl3Fe Molecular Weight (g/mol): 162.20 MDL Number: MFCD00011005 InChI Key: RBTARNINKXHZNM-UHFFFAOYSA-K IUPAC Name: iron(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Fe+3]
VeriSpec™ Iron Standard for ICP 1000 ppm in 2% HCl, Ricca Chemical
CAS: 7705-08-0 Molecular Formula: Cl3Fe Molecular Weight (g/mol): 162.20 MDL Number: MFCD00011005 InChI Key: RBTARNINKXHZNM-UHFFFAOYSA-K IUPAC Name: iron(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Fe+3]
VeriSpec™ Iron (Fe) Standard for AAS 1000 ppm in 2% HCl, Ricca Chemical
CAS: 7705-08-0 Molecular Formula: Cl3Fe Molecular Weight (g/mol): 162.20 MDL Number: MFCD00011005 InChI Key: RBTARNINKXHZNM-UHFFFAOYSA-K IUPAC Name: iron(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Fe+3]
Ferric Chloride, MP Biomedicals
CAS: 7705-08-0 Molecular Formula: Cl3Fe Molecular Weight (g/mol): 162.20 MDL Number: MFCD00011005 InChI Key: RBTARNINKXHZNM-UHFFFAOYSA-K Synonym: Chlorure Perrique,Iron (III) Chloride IUPAC Name: iron(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Fe+3]
Ferric Chloride, Anhydrous, Powder, Purified, Spectrum™ Chemical
CAS: 7705-08-0
Iron(III) chloride, MilliporeSigma™
CAS: 7705-08-0 Molecular Formula: Cl3Fe Molecular Weight (g/mol): 162.20 MDL Number: MFCD00011005 InChI Key: RBTARNINKXHZNM-UHFFFAOYSA-K Synonym: Ferric chloride; Iron trichloride IUPAC Name: iron(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Fe+3]