missing translation for 'onlineSavingsMsg'
Learn More

Ferric Chloride, 100% (w/v) (100 g + 40 mL Water), Ricca Chemical

Catalog No. p-3251674
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
31304 120 mL Natural Poly Bottle
313016 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31304 Supplier Ricca Chemical Company Supplier No. 31304
Only null left

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Color Amber
Percent Purity 100%
Grade Laboratory
Packaging Natural Poly Bottle
pH Acidic
Quantity 120 mL
Physical Form Liquid
Concentration or Composition Notes 100% (w/v) Aqueous Solution (100g + 40mL Water)
Chemical Name or Material Ferric Chloride