missing translation for 'onlineSavingsMsg'
Learn More

Ferric Chloride, 0.025% (w/v) as Hexahydrate, for BOD, LabChem™

Catalog No. LC143002
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC143002 1 L
LC143001 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC143002 Supplier LabChem Supplier No. LC143002

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10025-77-1 , 7732-18-5
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Color Brown-Yellow
CAS Max % 99.975, 0.025
Linear Formula FeCl3·6H2O
Density 1g/mL
Solubility Information Soluble in water
Formula Weight 270.3
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Decomposition Information Hydrogen chloride; Iron oxide; Sulfur compounds
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Ferric Chloride, 0.025% (w/v) as Hexahydrate, for BOD
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature