missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferric Ammonium Sulfate (Ferric Alum), Saturated, Ricca Chemical
$349.00 - $349.00
Chemical Identifiers
| CAS | 7783-83-7 |
|---|---|
| Molecular Formula | FeH28NO20S2 |
| Molecular Weight (g/mol) | 482.176 |
| InChI Key | LCPUDZUWZDSKMX-UHFFFAOYSA-K |
| Synonym | ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g |
| PubChem CID | 197097 |
| IUPAC Name | azanium;iron(3+);disulfate;dodecahydrate |
| SMILES | [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] |
Chemical Identifiers
| 7783-83-7 | |
| 482.176 | |
| ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g | |
| azanium;iron(3+);disulfate;dodecahydrate |
| FeH28NO20S2 | |
| LCPUDZUWZDSKMX-UHFFFAOYSA-K | |
| 197097 | |
| [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] |
Specifications
| Amber | |
| 66.0,32.24 | |
| FeH28NO20S2 | |
| LCPUDZUWZDSKMX-UHFFFAOYSA-K | |
| azanium;iron(3+);disulfate;dodecahydrate | |
| 197097 | |
| Natural Poly Bottle | |
| ∼2.5 | |
| Liquid |
| 7783-83-7 , 7732-18-5 | |
| 68.0,33.56 | |
| ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g | |
| [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] | |
| 482.176 | |
| Laboratory | |
| Saturated Aqueous Solution | |
| 1 L | |
| Ferric Ammonium Sulfate, Ferric Alum |