missing translation for 'onlineSavingsMsg'
Learn More

Ferric Alum Indicator, Volhard, Ricca Chemical

Catalog No. p-3251662
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
30504 120 mL Natural Poly Bottle
305016 500 mL Natural Poly Bottle
305032 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 30504 Supplier Ricca Chemical Company Supplier No. RI30504
Only null left

Chemical Identifiers

CAS 7783-83-7 , 7697-37-2 , 7732-18-5
Molecular Formula FeH28NO20S2
Molecular Weight (g/mol) Mixture
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
PubChem CID 197097
IUPAC Name azanium;iron(3+);disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]

Specifications

Grade Indicator
Packaging Natural Poly Bottle
Color Amber
Quantity 120 mL
Physical Form Liquid
Chemical Name or Material Ferric Alum Indicator, Volhard