Learn More
United Scientific™ Fermentation Tubes
Catalog No. S000890
Change view
Click to view available options
Capacity (Metric):
5 mL
Catalog No. Capacity (Metric)
S000890 5 mL
1 options
Catalog No. S000890 Supplier United Scientific™ Supplier No. FERMTB5
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Fermentation Tubes for use with verifying and measuring gas production in fermentation experiments.
The process of fermentation is when microorganisms act upon organic materials and produce gas as a byproduct. Different microorganisms and different materials produce various and unique gases. These gases rise and need to be collected and measured. These fermentation tubes are made from durable borosilicate glass and include blue graduations. The open end has a bulb forholding the materials being fermented. As the gas produced by the microorganisms rises into the vertical tube, the graduated increments allow accurate measurement to intervals of 0.1 mL in the 5 mL tube, or 0.2 mL in the 20 mL tube. Both tubes have a round glass base.
  • Blue Graduations
  • Glass base holds the apparatus stable
  • Made from Borosilicate 3.3 glass
  • Available in multiple sizes
Specifications
Product Type Gassing Tube
Capacity (Metric) 5 mL