missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TopVision Low Melting Point Agarose

Catalog No. FERR0801
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR0801 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERR0801 Supplier Thermo Scientific™ Supplier No. R0801

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Perform in-gel enzymatic process experiments and nucleic acid recovery after electrophoresis with Thermo Scientific™ TopVision™ Low Melting Point Agarose.

Top Vision Low Melting Point Agarose is highly purified agarose with a low melting temperature certified by strict quality control test procedures. It is used for in-gel enzymatic process experiments and for nucleic acids recovery from gels using Agarase (#EO0461).

Thermo Scientific TopVision Low Melting Point Agarose is used for in-gel enzymatic process experiments and for nucleic acid recovery after electrophoresis.

Highlights

  • Optimal concentration between 0.7-2% in all typical buffer systems
  • GQ (Genetic Quality) certified, which ensures that nucleic acids recovered from preparative gels can be used for downstream applications (enzymatic reactions etc.)
  • Low DNA/RNA binding
  • Excellent gel transparency
  • DNase and RNase free
  • Suitable for RNA analysis

Applications

  • Analytical electrophoresis of nucleic acids
  • Preparative electrophoresis
  • In-gel enzymatic processing experiments
  • Preparation of DNA preparative gels in routine molecular biology techniques
  • The DNA recovered from agarose gels after electrophoresis can be used in enzymatic process (restriction, ligation, etc.)

Specifications

Separation Range 100 to >30000 bp
Melting Point 65.5°C
Quantity 25 g
Validated Application Gel Electrophoresis
Product Line TopVision
Product Type Agarose Gel

For Research Use Only. Not for use in diagnostic procedures.