missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Taq DNA Polymerase, recombinant (5 U/μL)

Catalog No. FEREP0401
Change view
Click to view available options
Quantity:
10 x 500 Units
100 Units
5 x 500 Units
500 Units
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEREP0401 100 Units
FEREP0402 500 Units
FEREP0405 5 x 500 Units
FEREP0406 10 x 500 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. FEREP0401 Supplier Thermo Scientific™ Supplier No. EP0401

Thermo Scientific Taq DNA Polymerase is a highly thermostable DNA polymerase from the thermophilic bacterium Thermus aquaticus.

Thermo Scientific Taq DNA Polymerase is a highly thermostable DNA polymerase from the thermophilic bacterium Thermus aquaticus. The enzyme catalyzes 5' to 3' synthesis of DNA, has no detectable 3' to 5' exonuclease (proofreading) activity, and possesses low 5' to 3' exonuclease activity. In addition, Taq DNA Polymerase exhibits deoxynucleotidyl transferase activity, which frequently results in the addition of extra adenines at the 3'-end of PCR products. Recombinant Taq DNA Polymerase is the ideal tool for standard PCR of templates 5 kb or shorter.

Features

  • Thermostable—half life is more than 40 min at 95°C
  • Generates PCR products with 3'-dA overhangs
  • Supplied with two buffers: 10X Taq Buffer with KCl and 10X Taq Buffer with (NH4)2SO4
  • Incorporates modified nucleotides (e.g., biotin-, digoxigenin-, fluorescent-labeled nucleotides)

Applications

  • Routine PCR amplification of DNA fragments up to 5 kb
  • High throughput PCR
  • DNA labeling

Notes

  • The error rate of Taq DNA Polymerase in PCR is 2.2 x 10-5 errors per nt per cycle. Accordingly, the accuracy of PCR is 4.5 x 104. Accuracy is an inverse of the error rate and shows an average number of correct nucleotides incorporated before an error occurs.
  • 10X Taq Buffer without Detergent is recommended for microarray experiments.
  • 10X Taq Buffer with (NH4)2SO4 allows for PCR at wide range of magnesium concentrations and decreases unspecific priming.

Specifications

Concentration 5 U/μL
Content And Storage Taq DNA Polymerase (5 U/μL), 20 μL
• 10X Taq Buffer with KCl, 0.6 mL
• 10X Taq Buffer with (NH4)2SO4, 0.6 mL
• 25 mM MgCl2, 0.6 mL

Store at -20°C.
GC-Rich PCR Performance Low
Polymerase Taq DNA Polymerase
Reaction Speed Standard
Product Type Taq DNA Polymerase (Recombinant)
Quantity 100 Units
Shipping Condition Dry Ice
For Use With (Application) Standard PCR
Fidelity (vs. Taq) 1X
Hot Start No
Overhang 3'-A
Reaction Format Separate Components
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.