missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ X-Gal Solution, ready-to-use

Catalog No. FERR0941
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR0941 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERR0941 Supplier Thermo Scientific™ Supplier No. R0941

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Effectively screen colonies with X-Gal, an inert chromogenic substrate for beta-galactosidase which hydrolyzes X-Gal, forming an intense blue precipitate.

Formula: C14H15BrClNO6 Molecular weight: 408.6

X-Gal (5-bromo-4-chloro-3-indolyl-beta-D-galacto-pyranoside) is an inert chromogenic substrate for beta-galactosidase which hydrolyzes X-Gal into colorless galactose and 4-chloro-3-brom-indigo, forming an intense blue precipitate. Induction of the lacZ gene with IPTG leads to the hydrolysis of X-Gal and to the development of blue colonies (see Supporting data).

X-Gal Solution, ready-to-use, is stable, 0.22 μm membrane filtered solution formulated for direct use in conjunction with IPTG for blue/white colony screening.

Induction of the lacZ gene with IPTG leads to the hydrolysis of X-Gal and to the development of blue colonies. X-Gal Solution, ready-to-use, is a stable, 0.22μm membrane filtered solution formulated for direct use in conjunction with IPTG for blue/white colony screening.

Recommended for:

Blue/white colony screening to distinguish recombinant (white) from non-recombinant (blue) colonies (1); Visualization of beta-galactosidase reporter gene expression in transfected eukaryotic cells; Detection of beta-galactosidase activity in immunological and histochemical procedures.

Note:

Preparation of a 20mg/mL stock solution in dimethylformamide (DMF) or dimethylsulfoxide (DMSO) is recommended for X-Gal. DMF dissolves some plastic materials. The direct addition of DMF-containing solution to plastic Petri dishes should be avoided. DMF is toxic. Allow X-Gal solution containing dimethylformamide to completely dry before plating bacteria on agar plates.

Specifications

Detection Method Colorimetric
Product Type X-Gal Solution
Quantity 10 mL
Substrate Properties Chemical Substrate
Substrate Type Beta-Gal Substrate, X-Gal

For Research Use Only. Not for use in diagnostic procedures.