missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ pUC19 DNA

Catalog No. FERSD0061
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERSD0061 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERSD0061 Supplier Thermo Scientific™ Supplier No. SD0061

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Optimize cloning with this double-stranded closed circular high copy plasmid DNA, 2686 base pairs with a molecular weight of 1.74 x 106 Da isolated from E.coli.

pUC18 and pUC19 vectors are small, high copy number, E. coli plasmids, 2686 bp in length. They are identical except that they contain multiple cloning sites (MCS) arranged in opposite orientations.

Highlights

  • Purified by chromatography using proprietary patented technology
  • More than 90% in the supercoiled form
  • Isolated from E.coli (dam+, dcm+)

Specifications

Cloning Method Restriction Enzyme⁄ MCS
Concentration 0.5 μg/μL
Sample Type DNA
Final Product Type DNA
For Use With (Application) Cloning
Product Type DNA
Quantity 50 μg
Vector pUC19

For Research Use Only. Not for use in diagnostic procedures.