missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Maxima SYBR Green/Fluorescein qPCR Master Mix (2X)

Ready-to-use solution optimized for qPCR and 2-step RT-qPCR.

$389.91 - $4,846.63

Specifications

Concentration 2X
Detection Method SYBR
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
View More Specs

Products 3
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
FERK0241
View Documents Promotion Details
Thermo Scientific™
K0241
200 reactions
Each for $389.91
Only null left
 
FERK0243
View Documents Promotion Details
Thermo Scientific™
K0243
4,000 Reactions
Each for $4,846.63
Only null left
 
FERK0242
View Documents Promotion Details
Thermo Scientific™
K0242
1,000 Reactions
Each for $1,649.64
Only null left
 
Description

Description

The master mixes include Maxima Hot Start Taq DNA Polymerase and dNTPs in an optimized PCR buffer. Only template and primers need to be added. The SYBR Green I intercalating dye allows for DNA detection and analysis without using sequence-specific probes.

Maxima Hot Start Taq DNA Polymerase in combination with an optimized buffer ensures PCR specificity and sensitivity. dUTP is included in the mix for optional carry-over contamination control using uracil DNA glycosylase (UDG). The use of Maxima SYBR Green qPCR Master Mixes in real-time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral, and cDNA templates. Maxima SYBR Green qPCR Master Mixes are compatible with most real-time thermal cyclers.

  • Specificity - Maxima Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
  • Sensitivity - detects low copy number targets
  • Wide linear range - accurate quantification across 9 orders of magnitude
  • Reproducibility and convenience - ready-to-use 2X master mix
Specifications

Specifications

2X
Tube
qPCR
Standard
Maxima
Gene Expression
SYBR
High
Taq DNA Polymerase
BioRad iCycler iQ, BioRad MyiQ, BioRad iQ5
Real Time PCR SYBR Master Mix
DNA, dsDNA
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.