Learn More
Thermo Scientific™ Maxima SYBR Green qPCR Master Mix (2X), with separate ROX vial
Ready-to-use solution optimized for qPCR and 2-step RT-qPCR.
$385.63 - $4,835.56
Specifications
| Concentration | 2X |
|---|---|
| Detection Method | SYBR |
| Format | Tube |
| GC-Rich PCR Performance | High |
| PCR Method | qPCR |
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
|
FERK0251
|
Thermo Scientific™
K0251 |
200 reactions |
Each for $385.63
|
|
|||||
|
FERK0253
|
Thermo Scientific™
K0253 |
4,000 Reactions |
Each for $4,835.56
|
|
|||||
|
FERK0252
|
Thermo Scientific™
K0252 |
1,000 Reactions |
Each for $1,647.50
|
|
|||||
Description
The master mixes include Maxima Hot Start Taq DNA Polymerase and dNTPs in an optimized PCR buffer. Only template and primers need to be added. The SYBR Green I intercalating dye allows for DNA detection and analysis without using sequence-specific probes.
Maxima Hot Start Taq DNA Polymerase in combination with an optimized buffer ensures PCR specificity and sensitivity. dUTP is included in the mix for optional carry-over contamination control using uracil DNA glycosylase (UDG). The use of Maxima SYBR Green qPCR Master Mixes in real-time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral, and cDNA templates. Maxima SYBR Green qPCR Master Mixes are compatible with most real-time thermal cyclers.
- Specificity - Maxima Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
- Sensitivity - detects low copy number targets
- Wide linear range - accurate quantification across 9 orders of magnitude
- Reproducibility and convenience - ready-to-use 2X master mix
Specifications
| 2X | |
| Tube | |
| qPCR | |
| Standard | |
| Maxima | |
| Gene Expression |
| SYBR | |
| High | |
| Taq DNA Polymerase | |
| BioRad CFX96™, MJ Chromo4, MJ Opticon, Eppendorf MasterCycler RealPlex, Bio-Rad CFX384 Touch™, Roche LightCycler 480, Cepheid SmartCycler, Corbett RotorGene | |
| Real Time PCR SYBR Master Mix | |
| DNA, dsDNA |
For Research Use Only. Not for use in diagnostic procedures.