missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Maxima SYBR Green qPCR Master Mix (2X), with separate ROX vial

Ready-to-use solution optimized for qPCR and 2-step RT-qPCR.

Supplier:  Thermo Scientific™ K0251

 View more versions of this product

Catalog No. FERK0251


Only null left

Description

Description

The master mixes include Maxima Hot Start Taq DNA Polymerase and dNTPs in an optimized PCR buffer. Only template and primers need to be added. The SYBR Green I intercalating dye allows for DNA detection and analysis without using sequence-specific probes.

Maxima Hot Start Taq DNA Polymerase in combination with an optimized buffer ensures PCR specificity and sensitivity. dUTP is included in the mix for optional carry-over contamination control using uracil DNA glycosylase (UDG). The use of Maxima SYBR Green qPCR Master Mixes in real-time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral, and cDNA templates. Maxima SYBR Green qPCR Master Mixes are compatible with most real-time thermal cyclers.

  • Specificity - Maxima Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
  • Sensitivity - detects low copy number targets
  • Wide linear range - accurate quantification across 9 orders of magnitude
  • Reproducibility and convenience - ready-to-use 2X master mix
Specifications

Chemical Identifiers

2X
Tube
qPCR
Standard
Maxima
Gene Expression
SYBR
High
Taq DNA Polymerase
BioRad CFX96™, MJ Chromo4, MJ Opticon, Eppendorf MasterCycler RealPlex, Bio-Rad CFX384 Touch™, Roche LightCycler 480, Cepheid SmartCycler, Corbett RotorGene
Real Time PCR SYBR Master Mix
DNA, dsDNA

Specifications

2X
SYBR
High
Taq DNA Polymerase
BioRad CFX96™, MJ Chromo4, MJ Opticon, Eppendorf MasterCycler RealPlex, Bio-Rad CFX384 Touch™, Roche LightCycler 480, Cepheid SmartCycler, Corbett RotorGene
Real Time PCR SYBR Master Mix
200 Reactions of 25 μL
200 reactions
2 x 1.25 mL
Contains:
• 2 x 1.25 mL Maxima SYBR Green qPCR Master Mix (2X) containing Maxima Hot Start Taq DNA Polymerase, dNTPs (also dUTP) and SYBR Green I in an optimized PCR buffer; sufficient for 200 x 25 μL reactions.
• 50 μL ROX solution, 50 μM
• 2 x 1.25 mL Nuclease-Free Water

Store at -20°C.
Tube
qPCR
Standard
Maxima
200 reactions
Gene Expression
DNA, dsDNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.