Learn More
Thermo Scientific™ Maxima Probe qPCR Master Mix (2X), with separate ROX vial
Optimize qPCR with probe chemistry using standard cycling protocols with these ready-to-use qPCR master mixes.
$399.55
Specifications
| Concentration | 2X |
|---|---|
| Detection Method | Primer-probe |
| Format | Tube |
| GC-Rich PCR Performance | High |
| PCR Method | qPCR |
Description
Maxima Probe qPCR Master Mixes are ready-to-use solutions optimized for quantitative real-time PCR. The master mixes include Maxima Hot Start Taq DNA Polymerase and dNTPs in an optimized PCR buffer. Only template and primers need to be added.
Maxima Hot Start Taq DNA Polymerase in combination with an optimized buffer ensures PCR specificity and sensitivity. dUTP is included in the mix for optional carry-over contamination control using uracil DNA glycosylase (UDG). The use of Maxima Probe qPCR Master Mixes in real-time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral, and cDNA templates.
Maxima Probe qPCR Master Mixes are compatible with most real-time thermal cyclers.
Highlights
- Specificity - Maxima Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
- Sensitivity - detects low copy number targets
- Wide linear range - accurate quantification across 9 orders of magnitude
- Reproducibility and convenience - ready-to-use 2X master mix
Specifications
| 2X | |
| Tube | |
| qPCR | |
| Standard | |
| Maxima | |
| Gene Expression |
| Primer-probe | |
| High | |
| Taq DNA Polymerase | |
| BioRad CFX96™, BioRad iCycler iQ, BioRad iQ5, BioRad MyiQ, MJ Chromo4, Eppendorf MasterCycler RealPlex, Corbett RotorGene, Bio-Rad CFX384 Touch™, Roche LightCycler 480, Cepheid SmartCycler, MJ Opticon | |
| qPCR Master Mix | |
| DNA, dsDNA |
For Research Use Only. Not for use in diagnostic procedures.