missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET Plasmid Maxiprep Kit

Numéro de catalogue. FERK0491
Modifier l'affichage
Click to view available options
No. of Reactions:
10 Preps
25 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. No. of Reactions
FERK0491 10 Preps
FERK0492 25 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. FERK0491 Fournisseur Thermo Scientific™ Code fournisseur K0491
Il en reste null

Thermo Scientific GeneJET Plasmid Maxiprep Kit, sufficient for 25 preps, utilizes silica-based membrane technology in the form of convenient spin column.

Thermo Scientific GeneJET Plasmid Maxiprep Kit, sufficient for 25 preps, utilizes silica-based membrane technology in the form of convenient spin column. Each prep recovers greater than 400 ng/μL of high copy plasmid DNA that can be used in a wide variety of molecular biology procedures, such as restriction endonuclease digestion, PCR, cloning, transformation, automated sequencing, in vitro transcription, and transfection of robust cell lines.

Highlights

  • Efficient—high yields up to 750 μg of high quality plasmid DNA from 250 mL of bacterial culture
  • High concentration of DNA purified— >400 ng/μL
  • Convenient—spin column format, centrifuge or vacuum protocols
  • Fast—procedure takes 60 minutes, considerably faster than using gravity flow columns

Applications

Isolated plasmid DNA is suitable for all conventional molecular biology procedures, including:

  • Transfection of robust cell lines
  • FastDigest restriction enzyme digestion
  • Automated fluorescent sequencing
  • PCR
  • Cloning
  • Transformation
  • In vitro transcription

Spécifications

Concentration 400 ng/μL
Column Type Spin Column
Format Spin Column
Isolation Technology Silica Column
Scale Maxi
Sample Type Bacteria
For Use With (Application) Real-Time Quantitative PCR (qPCR), Transfection, Cloning, Sequencing, In Vitro Transcription, Transformation, Nucleic Acid Labeling, PCR
High-throughput Compatibility Not High-throughput Compatible (Manual)
No. of Reactions 10 Preps
Prep Scale > 200 μg (Large-Scale) Plasmid DNA
Product Line GeneJET
Product Type Plasmid MaxiPrep Kit
Quantity 10 preps
Target Plasmid DNA
Test Time 60 min.
Afficher plus de résultats Afficher moins de résultats

For Research Use Only. Not for use in diagnostic procedures.