missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET Plant Genomic DNA Purification Kit

Catalog No. FERK0792
Change view
Click to view available options
Quantity:
250 Preps
50 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERK0792 250 Preps
FERK0791 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERK0792 Supplier Thermo Scientific™ Supplier No. K0792

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific GeneJET Plant Genomic DNA Purification Mini Kit is designed for rapid and efficient purification of high quality genomic DNA from a wide variety of plant species and tissue types.

GeneJET Plant Genomic DNA Purification Mini Kit is designed for rapid and efficient purification of high quality genomic DNA from a wide variety of plant species and tissue types. It utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation.

Thermo Scientific GeneJET Plant Genomic DNA Purification Mini Kit is designed for rapid and efficient purification of high quality genomic DNA from a wide variety of plant species and tissue types. It utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation.

Highlights
• Fast—procedure completed in 30 min following cell lysis
• Convenient—protocol does not include clogging-prone lysate filtration step
• High yield—up to 32 μg of high quality DNA from 100 mg sample, including nuclear, mitochondrial, and chloroplast DNA
• Compatible—use with awide variety of samples (leaves, roots, sprouts and other parts of various plants)

The standard procedure takes less than 30 minutes following cell lysis and yields purified DNA greater than 30 kb in size. DNA yields vary between different species and tissues depending on genome size, ploidy, cell number, and the age of the tissue sample. The typical yield from a standard tissue source (e. g. young wheat leaves) is 30 to 32 μg from 100 mg of tissue. Isolated DNA can be used directly in PCR, qPCR, southern blotting, and enzymatic reactions.

Applications
• Isolated genomic DNA is ideal for use in common molecular biology procedures, including:
• PCR and qPCR
• FastDigest and conventional restriction digestion
• Southern blotting

Specifications

Column Type Spin Column
Content And Storage • 6 x 1 mL RNase A; store at room temperature until opened, –20°C after opening
• 120 mL Lysis Buffer A; room temperature
• 15 mL Lysis Buffer B; room temperature
• 40 mL Precipitation Solution; room temperature
• 120 mL Plant gDNA Binding Solution; room temperature
• 40 mL Wash Buffer I (concentrated); room temperature
• 40 mL Wash Buffer II (concentrated); room temperature
• 150 mL Elution Buffer; room temperature
• 250 GeneJET Purification Columns with Collection Tubes; room temperature
• 250 Collection Tubes; room temperature

Isolation Technology Silica Spin Column
Scale Mini
Sample Type Plants
Elution Volume 200 μL
Final Product Type gDNA
For Use With (Application) PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization
High-throughput Compatibility Not High-throughput Compatible (Manual)
Quantity 250 Preps
Shipping Condition Room Temperature
Test Time 30 min. (After sample homogenization)
Yield ≤32 μg per 100 mg tissue
Starting Material Amount ≤100 mg of fresh/frozen
≤20 mg lyophilized
Show More Show Less
Can I use water instead of Elution Buffer in the elution step of GeneJET and MagJET nucleic acid purification kits?

We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.

Can I use a vacuum manifold with GeneJET kits?

The GeneJET spin columns are compatible with vacuum manifolds of standard size (e.g. Qiagen, Sigma-Aldrich, Promega). However, not all GeneJET kits are supported with a vacuum protocol. Please refer to the particular kit protocol for the detailed procedure.

At what temperature should I perform centrifugation steps in the GeneJET kits?

We recommend performing centrifugation steps at room temperature when the temperature is not specified.

Are GeneJET spin columns or MagJET magnetic beads available separately?

No, we do not offer GeneJET columns or MagJET magnetic beads separately at this time.

For Research Use Only. Not for use in diagnostic procedures.