missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Fast DNA End Repair Kit

Numéro de catalogue. ferk0771
Change view
Click to view available options
Quantity:
50 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
FERK0771 50 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. FERK0771 Fournisseur Thermo Scientific™ Code fournisseur K0771
Il en reste null

Perform blunting and phosphorylation of DNA ends in just 5 minutes for subsequent use in blunt-end ligation with this kit.

Fast DNA End Repair Kit is used for blunting and phosphorylation of DNA ends in just 5 minutes for subsequent use in blunt-end ligation.

All components of the kit contain premixed reagents to reduce pipetting steps and provide convenience. The End Repair Enzyme Mix contains an optimized mixture of T4 DNA Polymerase and Klenow Fragment to achieve highly effective blunting of fragmented DNA, and T4 Polynucleotide Kinase (PKN) for efficient phosphorylation of DNA ends. The 10X End Repair Reaction Mix contains an optimized reaction buffer, ATP, and dNTPs.

Samples such as fragmented genomic DNA (restriction enzyme digested, nebulized or sonicated), restriction enzyme digested plasmid DNA, double stranded cDNA, and PCR products containing dA overhangs are all compatible with the kit.

Highlights

During the DNA end repair reaction, fragmented DNA is converted into blunt-end DNA containing a 5'-phosphate and 3'-hydroxyl groups. The 5'→3' polymerase activity of the End Repair Enzyme Mix fills-in 5' protruded DNA ends while 3'→5' exonuclease activity removes 3'-overhangs. T4 PNK adds 5'-phosphates to ends of unphosphorylated DNA fragments, such as PCR products.

Highlights

  • Efficient - blunting and phosphorylation of 0.5 to 5 µg DNA
  • Fast -reaction is completed in 5 minutes
  • Convenient -reaction components are premixed to reduce pipetting steps

Spécifications

Format Kit
Sample Type DNA
Enzyme DNA Polymerase, T4 Polynucleotide Kinase
For Use With (Application) Cloning
Product Type Fast DNA End Repair Kit
Quantity 50 Reactions

For Research Use Only. Not for use in diagnostic procedures.