missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DTT (dithiothreitol)

Catalog No. FERR0862
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR0862 25 g
FERR0861 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERR0862 Supplier Thermo Scientific™ Supplier No. R0862

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Stabilize enzymes and other proteins which possess free sulfhydryl groups with DTT, shown to restore activity lost by oxidation of these groups in vitro.

DTT (DL-Dithiothreitol; Clelands reagent) is used to stabilize enzymes and other proteins which possess free sulfhydryl groups. It has been shown to restore activity lost by oxidation of these groups in vitro. DTT quantitatively reduces disulfide bonds and maintains monothiols in a reduced state .

At a final 0.1 M concentration, DTT is also widely used for disruption of protein disulfide bonds in SDS-polyacrylamide gel electrophoresis.

DTT quantitatively reduces disulfide bonds and maintains monothiols in a reduced state (1). At a final 0.1M concentration DTT is also widely used for disruption of protein disulfide bonds in SDS-polyacrylamide gel electrophoresis.
  • Free of endo-, exodeoxyribonucleases, ribonucleases and phosphatases

Recommended for:

Enzyme stabilization; Enzymatic reactions; Protein electrophoresis.

Note:

DTT is not stable in solution. Only freshly-made DTT solutions should be used. To prepare a 1M DTT solution, dissolve 1.55g of DTT powder in 10mL of deionized water (e.g., Water, nuclease-free).

Specifications

Concentration 0.1 M
Product Type DTT (Dithiothreitol)
Quantity 25 g
How do I prepare a 1 M DTT solution from powder and how should I store the solution?

Preparing 1 M DTT:
1) Dissolve 3.09 g DTT in 20 mL distilled H2O.
2) Dispense into 1 mL aliquots.
3) Store the aliquots at -20 degrees C.
Note: Upon thawing the vial, there may be some precipitation that can be re-dissolved by warming and mixing.

For Research Use Only. Not for use in diagnostic procedures.