missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ dITP Solution (100 mM)

Catalog No. FERR1191
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR1191 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERR1191 Supplier Thermo Scientific™ Supplier No. R1191

Thermo Scientific dITP (2'-deoxyinosine 5'-triphosphate) is a highly stable nucleotide supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH.

Thermo Scientific dITP (2'-deoxyinosine 5'-triphosphate) is a highly stable nucleotide supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH. The nucleotide has greater than 99% purity and is free of nuclease activities and human, and E. coli DNA. Thermo Scientific dITP is designed for use with a wide variety of different molecular biology applications.

Applications
• PCR and RT-PCR with Taq DNA polymerase (Cat. No. EP0072)
• qPCR
• cDNA synthesis
• SNP genotyping
• Primer extension.

Note: dITP is not suitable for PCR with proofreading enzymes or enzyme mixes containing proofreading enzymes.

Specifications

Concentration 100 mM
Content And Storage dITP Solution, 100 mM (0.25 mL)

Store at –20°C.
Form Liquid
Polymerase DNA Polymerase
Label or Dye Unlabeled
Quantity 0.25 mL
Shipping Condition Dry Ice
For Use With (Application) Standard PCR, cDNA Synthesis, RT-PCR

For Research Use Only. Not for use in diagnostic procedures.