missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CpG Methylated Human Genomic DNA

Catalog No. FERSD1131
Change view
Click to view available options
Quantity:
15 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERSD1131 15 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERSD1131 Supplier Thermo Scientific™ Supplier No. SD1131

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ensure accurracy in DNA methylation analysis with this methylated human genomic DNA control.

Human genomic DNA is enzymatically methylated with CpG Methyltransferase (M.SssI). Genomic DNA was isolated from blood samples from healthy male (HIV, HBV, and HCV negative) individuals. Isolated genomic DNA was treated with CpG Methyltransferase, repurified, and dissolved in TE buffer. = 98% CpG methylation level confirmed by bisulfite sequencing.

Highlights:

  • Pure—DNA free from inhibitors, proteins and RNA
  • Convenient—ready to use 100 ng/μL concentration
  • Compatible with downstream applications
  • Complete methylation of cytosines in CpG dinucleotides

Specifications

Concentration 100 ng/μL
Format Tube
For Use With (Application) Sequencing
Includes [other]
Product Type CpG Methylated Human Genomic DNA
Quantity 15 μg
Species Human
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.