missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Biotin-11-dUTP Solution (1 mM)

Catalog No. FERR0081
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR0081 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERR0081 Supplier Thermo Scientific™ Supplier No. R0081

Thermo Scientific Biotin-11-dUTP (biotin-epsilon-aminocaproyl-[5-{3-aminoallyl}-2'-deoxyuridine-5'-triphosphate]) is supplied as 1 mM aqueous solution titrated to pH 7.0 with NaOH and is designed for enzymatic non-radioactive labelling of DNA.

Thermo Scientific Biotin-11-dUTP (biotin-epsilon-aminocaproyl-[5-{3-aminoallyl}-2'-deoxyuridine-5'-triphosphate]) is supplied as 1 mM aqueous solution titrated to pH 7.0 with NaOH and is designed for enzymatic non-radioactive labelling of DNA.

Applications
• Enzymatic non-radioactive labeling of DNA by PCR
• Nick-translation
• cDNA synthesis
• Random primed labeling
• Primer extension

Biotin-11-dUTP can be enzymatically incorporated into DNA with:
• Reverse transcriptases
Taq DNA Polymerase (Cat. No. EP0072)
• phi29 DNA Polymerase (EP0091)
• Klenow Fragment, exo (EP0421)
• Klenow Fragment (EP0051)
• DNA Polymerase I (EP0041)

Specifications

Concentration 1 mM
Content And Storage Biotin-11-dUTP Solution, 1 mM (50 μL)

Store at –20°C.
Form Liquid
Polymerase DNA Polymerase
Labeling Method Direct Labeling
Label or Dye Biotin
Quantity 50 μL
Shipping Condition Dry Ice
For Use With (Application) cDNA Synthesis, Non-Radioactive Labeling

For Research Use Only. Not for use in diagnostic procedures.