Learn More
Thermo Scientific™ AarI-AjiI-Bpu10I-ScaI-PasI Buffer (10X)
Optimize digestions with AarI, AjiI, Bpu10I, ScaI, and PasI restriction enzymes using this buffer.
Supplier: Thermo Scientific™ B27
Description
To ensure consistent enzyme performance, Thermo Scientific restriction enzyme buffers contain BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations. Multiple freeze-thaw cycles of the buffers will not cause BSA precipitation.
restriction enzymes exhibit 100% of their certified activity in the recommended buffer.
The following enzymes require unique buffers for optimal digestion:Â
- AarIÂ
- AjiIÂ
- BamHIÂ
- BfuIÂ
- Bpu10IÂ
- BseXIÂ
- Bsp143IÂ
- Cfr9IÂ
- Cfr10IÂ
- Eam1105IÂ
- Ecl136IIÂ
- Eco52IÂ
- EcoRIÂ
- KpnIÂ
- Lsp1109IÂ
- PacIÂ
- PasIÂ
- Ppu21IÂ
- SacIÂ
- ScaIÂ
- SdaIÂ
- SduIÂ
- SgeIÂ
- TaqIÂ
Specifications
| 10X | |
| 1 mL | |
| Traditional Cloning |
| AarI, AjiI, Bpu10I, ScaI, PasI | |
| Buffer |
For Research Use Only. Not for use in diagnostic procedures.