missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fenofibrate, 98%

Catalog No. AC461780250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461780250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC461780250 Supplier Thermo Scientific Chemicals Supplier No. 461780250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 49562-28-9 | C20H21ClO4 | 360.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 49562-28-9
Molecular Formula C20H21ClO4
Molecular Weight (g/mol) 360.83
InChI Key YMTINGFKWWXKFG-UHFFFAOYSA-N
PubChem CID 3339
ChEBI CHEBI:5001

Specifications

Melting Point 79°C to 82°C
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether,soluble in acetone,,soluble in ethyl acetate,soluble in acetonitrile,soluble in methanol,soluble in ethanol
Formula Weight 360.83
Percent Purity 98%
Chemical Name or Material Fenofibrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
May cause long lasting harmful effects to aquatic life.
Precautionary Statement GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical attention/advice if you feel unwell.
Avoid release to the environment.
Dispose of contents/container to an approved waste disposal plant.
EINECSNumber 256-376-3

RUO – Research Use Only