missing translation for 'onlineSavingsMsg'
Learn More

(+)-Fenchone 98.0+%, TCI America™

Catalog No. F016325ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F016325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F016325ML Supplier TCI America Supplier No. F016325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4695-62-9
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.237
MDL Number MFCD00070689
InChI Key LHXDLQBQYFFVNW-XCBNKYQSSA-N
Synonym +-fenchone, 1s-1,3,3-trimethylbicyclo 2.2.1 heptan-2-one, 1s-1,3,3-trimethyl-norbornan-2-one
PubChem CID 1201521
ChEBI CHEBI:165
IUPAC Name (1R,4S)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one
SMILES CC1(C2CCC(C2)(C1=O)C)C

Specifications

Color Yellow
Boiling Point 193°C
Quantity 25 mL
Formula Weight 152.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (+)-Fenchone
EINECSNumber (4)-1978
RTECSNumber RB7875200
TSCA Yes