missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Febuxostat

Catalog No. 442710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
442710 10 mg
442750 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 442710 Supplier Tocris Bioscience™ Supplier No. 4427/10

Xanthine oxidase inhibitor

Non-purine inhibitor of xanthine oxidase (Ki = 1.2 nM). Binds to a channel leading to the enzyme active site; obstructs substrate binding.

Chemical Identifiers

CAS 144060-53-7
Molecular Formula C16H16N2O3S

Specifications

Quantity 10 mg
Formula Weight 316.37
Purity >99%
Molecular Weight (g/mol) 316.38
Chemical Name or Material Febuxostat
Recommended Storage Store at Room Temperature