missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FD and C Yellow No. 5, Spectrum™ Chemical
FD150, 1934-21-0, C16H9N4O9S2Na3
$343.82 - $18,679.96
Chemical Identifiers
| CAS | 1934-21-0 |
|---|---|
| Molecular Formula | C16H9N4Na3O9S2 |
| Molecular Weight (g/mol) | 534.36 |
| InChI Key | UJMBCXLDXJUMFB-UHFFFAOYNA-K |
| IUPAC Name | trisodium 5-oxo-1-(4-sulfonatophenyl)-4-[2-(4-sulfonatophenyl)diazen-1-yl]-4,5-dihydro-1H-pyrazole-3-carboxylate |
| SMILES | [Na+].[Na+].[Na+].[O-]C(=O)C1=NN(C(=O)C1N=NC1=CC=C(C=C1)S([O-])(=O)=O)C1=CC=C(C=C1)S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18602636
|
N/A | 100 g | Amber Glass Bottle |
Each for $343.82
|
|
||||
|
18602637
|
N/A | 500 g | Amber Glass Bottle |
Each for $937.21
|
|
||||
|
18602638
|
N/A | 2.5 kg | Amber Glass Bottle |
Each for $2,361.50
|
|
||||
|
18602639
|
N/A | 20 kg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18602640
|
N/A | 25 kg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical FD and C Yellow No. 5Chemical Identifiers
| 1934-21-0 | |
| 534.36 | |
| trisodium 5-oxo-1-(4-sulfonatophenyl)-4-[2-(4-sulfonatophenyl)diazen-1-yl]-4,5-dihydro-1H-pyrazole-3-carboxylate |
| C16H9N4Na3O9S2 | |
| UJMBCXLDXJUMFB-UHFFFAOYNA-K | |
| [Na+].[Na+].[Na+].[O-]C(=O)C1=NN(C(=O)C1N=NC1=CC=C(C=C1)S([O-])(=O)=O)C1=CC=C(C=C1)S([O-])(=O)=O |
Specifications
| 1934-21-0 | |
| C16H9N4Na3O9S2 | |
| [Na+].[Na+].[Na+].[O-]C(=O)C1=NN(C(=O)C1N=NC1=CC=C(C=C1)S([O-])(=O)=O)C1=CC=C(C=C1)S([O-])(=O)=O | |
| 534.36 | |
| Reagent | |
| FD and C Yellow No. 5 |
| 100% | |
| UJMBCXLDXJUMFB-UHFFFAOYNA-K | |
| trisodium 5-oxo-1-(4-sulfonatophenyl)-4-[2-(4-sulfonatophenyl)diazen-1-yl]-4,5-dihydro-1H-pyrazole-3-carboxylate | |
| 87% | |
| Orange |