missing translation for 'onlineSavingsMsg'
Learn More

FD and C Yellow No. 5, Spectrum™ Chemical

Catalog No. 18602636
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
20 kg
25 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602636 100 g Amber Glass Bottle
18602637 500 g Amber Glass Bottle
18602638 2.5 kg Amber Glass Bottle
18602639 20 kg Amber Glass Bottle
18602640 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18602636 Supplier Spectrum Chemical Mfg Cor Supplier No. FD150100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

FD150, 1934-21-0, C16H9N4O9S2Na3

Spectrum™ Chemical FD and C Yellow No. 5

Chemical Identifiers

CAS 1934-21-0
Molecular Formula C16H9N4Na3O9S2
Molecular Weight (g/mol) 534.36
InChI Key UJMBCXLDXJUMFB-UHFFFAOYNA-K
IUPAC Name trisodium 5-oxo-1-(4-sulfonatophenyl)-4-[2-(4-sulfonatophenyl)diazen-1-yl]-4,5-dihydro-1H-pyrazole-3-carboxylate
SMILES [Na+].[Na+].[Na+].[O-]C(=O)C1=NN(C(=O)C1N=NC1=CC=C(C=C1)S([O-])(=O)=O)C1=CC=C(C=C1)S([O-])(=O)=O

Specifications

CAS Max % 100%
Percent Purity 87%
Grade Reagent
Packaging Amber Glass Bottle
Color Orange
Quantity 100 g
Chemical Name or Material FD and C Yellow No. 5