missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FD and C Yellow No. 5, Spectrum™ Chemical
FD150, 1934-21-0, C16H9N4O9S2Na3
Supplier: Spectrum Chemical Mfg Cor FD15020KGBL
| Quantity | 20 kg |
|---|---|
| Packaging | Amber Glass Bottle |
Description
Spectrum™ Chemical FD and C Yellow No. 5Chemical Identifiers
| 1934-21-0 | |
| 534.36 | |
| trisodium 5-oxo-1-(4-sulfonatophenyl)-4-[2-(4-sulfonatophenyl)diazen-1-yl]-4,5-dihydro-1H-pyrazole-3-carboxylate |
| C16H9N4Na3O9S2 | |
| UJMBCXLDXJUMFB-UHFFFAOYNA-K | |
| [Na+].[Na+].[Na+].[O-]C(=O)C1=NN(C(=O)C1N=NC1=CC=C(C=C1)S([O-])(=O)=O)C1=CC=C(C=C1)S([O-])(=O)=O |
Specifications
| FD and C Yellow No. 5 | |
| 100% | |
| UJMBCXLDXJUMFB-UHFFFAOYNA-K | |
| trisodium 5-oxo-1-(4-sulfonatophenyl)-4-[2-(4-sulfonatophenyl)diazen-1-yl]-4,5-dihydro-1H-pyrazole-3-carboxylate | |
| 87% | |
| Amber Glass Bottle | |
| 20 kg |
| 1934-21-0 | |
| C16H9N4Na3O9S2 | |
| [Na+].[Na+].[Na+].[O-]C(=O)C1=NN(C(=O)C1N=NC1=CC=C(C=C1)S([O-])(=O)=O)C1=CC=C(C=C1)S([O-])(=O)=O | |
| 534.36 | |
| Reagent | |
| Orange |