missing translation for 'onlineSavingsMsg'
Learn More

FD and C Blue No. 1, Spectrum™ Chemical

Catalog No. 18602610
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
10 kg
45 kg
Packaging:
Fiber Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602610 25 g Fiber Drum
18602606 100 g Fiber Drum
18602607 500 g Fiber Drum
18602609 10 kg Fiber Drum
18602608 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18602610 Supplier Spectrum Chemical Mfg Cor Supplier No. FD11025GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

FD110, 3844-45-9, C37H34N2O9S3Na2

Spectrum™ Chemical FD and C Blue No. 1 is a synthetic dye produced from petroleum. It is used as a coloring agent, biological stain, textile dye, wood stain, and indicator. In cosmetics and personal care products, it is used in the formulation of a wide variety of product types, eye makeup, makeup, lipsticks, nail polishes, personal cleansers and skin care products. It appears as a dark violet to reddish violet, crystalline powder that is water soluble. In the United States, the Food and Drug Administration (FDA) controls the specifications for food, drug and cosmetic colors. Spectrum certifies all batches of artificial food colors meet FDA FD&C quality.

Chemical Identifiers

CAS 3844-45-9
Molecular Formula C37H34N2Na2O9S3
Molecular Weight (g/mol) 792.84
InChI Key SGHZXLIDFTYFHQ-UHFFFAOYSA-L
IUPAC Name disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(2-sulfonatophenyl)methylium
SMILES [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=CC=C1S([O-])(=O)=O

Specifications

CAS Max % 100%
Packaging Fiber Drum
Color Purple/Violet
Quantity 25 g
Percent Purity 85%
Grade Reagent
Chemical Name or Material FD and C Blue No. 1