missing translation for 'onlineSavingsMsg'
Learn More

Fatty Acids Unsaturated Kit (individualy packaged in quantities indicated), MilliporeSigma™ Supelco™

Catalog No. 111016472
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016472 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016472 Supplier Merck Emd Millipore Supplier No. UN101KT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Food and Beverages

Specifications

CAS 112-80-1
Shipping Condition Dry Ice
Physical Form Neat
Type Analytical Standard
Quantity 1 Ea.
Grade Analytical Standard