Learn More
Invitrogen™ ViewRNA™ Fast Red Buffer and Substrate Kit
Catalog No. QVT0504
Change view
Click to view available options
Quantity:
24 assays
Catalog No. Quantity
QVT0504 24 assays
1 options
Catalog No. QVT0504 Supplier Invitrogen™ Supplier No. QVT0504
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
ViewRNA Fast Red Buffer and Substrate Kit is an individual component of the ViewRNA Chromogenic Signal Amplification Kit.
ViewRNA Fast Red Buffer and Substrate Kit is an individual component of the ViewRNA Chromogenic Signal Amplification Kit, 24 assays (Cat. No. QVT0200). It is a red substrate used for the detection of alkaline phosphatase activity.
Specifications
Format Kit
Product Type RNA In Situ Hybridization Buffer and Substrate Kit
Quantity 24 assays
For Research Use Only. Not for use in diagnostic procedures.