missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Fast MicroSEQ™ 500 16S rDNA PCR Kit

Numéro de catalogue. 4370489
Encompass
Modifier l'affichage
Click to view available options
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue.
4370489 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 4370489 Fournisseur Applied Biosystems™ Code fournisseur 4370489

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

The Fast MicroSEQ™ 500 16S rDNA PCR kit utilizes new Fast technology and provides bacterial identifications from colony to results in as little as 5 hours using the Veriti™ 96-well Thermal Cycler.

The Fast MicroSEQ™ 500 16S rDNA PCR kit utilizes new Fast technology and provides bacterial identifications from colony to results in as little as 5 hours using the Veriti™ 96-well Thermal Cycler. This kit is also compatible with the GeneAmp™ PCR System 9700. The PCR-amplified products that are generated using this kit can be sequenced using the MicroSEQ™ 500 16S rDNA Seqencing Kit. The two kits together permit the amplification and sequencing of 500 base pairs of the 16S rRNA gene. These sequences can then be compared to the MicroSEQ™ ID Analysis Software 16S rDNA 500 Library to obtain an identity match. This kit does not include a protocol or a Quick Reference Card.

  • Sequence-based measurement ensures you get reproducible results in under 24 hours.
  • Growth requirement-independent measurement enables you to identify bacteria using objective criteria.
  • Universal domain-level primers eliminate up-front optimization.
  • MicroSEQ™ ID Analysis Software ensures accurate species identification.
  • Customizable libraries give you the flexibility to create new libraries and search validated libraries that contain the most frequently encountered organisms.
  • Requires minimal training-same work flow as the MicroSEQ™ D2 LSU fungal kit.

The MicroSEQ™ ID System: A Complete, Integrated Solution

Streamlining every step in microbial identification, Applied Biosystems™ MicroSEQ™ System combines the advantages of MicroSEQ™ ID Software with easy PrepMan Ultra sample preparation reagents and protocols, MicroSEQ™ bacterial and fungal application kits, and industry-leading thermal cyclers and sequencing systems. To ensure optimal performance and system-wide integration, we develop and test all components of the system together.

Spécifications

Contenu et stockage Each kit contains enough PCR Master Mix to perform 50 PCR amplifications, 5 negative control assays, and 5 positive control assays. One tube of positive control E. coli DNA, and one of water (for use as a negative control) are also included.

Store at -15°C to -25°C upon reciept. After first use, store at 2°C to 8°C in a PCR Clean Room.
Méthode de détection Primer-probe
Performance de la PCR pour les régions riches en GC Low
Vitesse de réaction Fast
Technique SBT (Sequence-Based Typing)
À utiliser avec (équipement) Veriti Thermal Cycler
Bibliothèques Customizable, Bacterial Library (> 2000 species)
Gamme de produit MicroSEQ™
Type de produit rDNA PCR Kit
Quantité 1 kit
Shipping condition (Condition d'expédition) Dry Ice
Logiciel MicroSEQ™ ID Analysis Software
Cible 1st 500 bp of 16S rRNA gene
Classe d’organismes cibles Bacteria
À utiliser avec (application) Standard PCR
Type d’échantillon DNA (Genomic)
Afficher plus de résultats Afficher moins de résultats

For Research Use Only. Not for use in diagnostic procedures.