missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fast Garnet GBC Base 97%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Assay Percent Range | 97% |
| UV Spectrometry | Complies |
| Linear Formula | NH2C6H3(CH3)N=NC6H4CH3 |
| Beilstein | 16, 344 |
| Merck Index | 15, 415 |
| Color | Brown/Red |
| Quantity | 25 g |
| Physical Form | Crystalline Powder |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement: May cause an allergic skin reaction. May cause cancer. |
| Precautionary Statement | GHS P Statement: Wear eye protection/face protection. Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 202-591-2 |
| RTECSNumber | XU8800000 |
| TSCA | TSCA |
RUO – Research Use Only