Learn More
Invitrogen™ Fast Blue Substrate (20 mL Blue Buffer + 420 uL each Blue Reagents 1-3)
Catalog No. QVT0506
Change view
Click to view available options
Quantity:
24 Assays
Catalog No. Quantity
QVT0506 24 Assays
1 options
Catalog No. QVT0506 Supplier Invitrogen™ Supplier No. QVT0506
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Fast Blue Substrate (20 mL Blue Buffer + 420 uL each Blue Reagents 1-3) is an individual component of the ViewRNA™ ISH Tissue Assay Kits, 2-Plex
Fast Blue Substrate (20 mL Blue Buffer + 420 uL each Blue Reagents 1-3) is an individual component of the ViewRNA™ ISH Tissue Assay Kits, 2-Plex, 24 assays (Cat. No. QVT0012). It is a blue precipitating substrate with components 1 -3 used for the detection of alkaline phosphatase activity.
Specifications
Detection Method Chromogenic, Fluorescence
Product Type Substrate
Quantity 24 Assays
Does ViewRNA Tissue Assay Blue Module (Cat Nos. 19932, 19943) use the same Fast Blue reagent as used in the earlier Fast Blue Substrate products?
Yes. There was no change to the Fast Blue reagent used.
How is ViewRNA Tissue Assay Blue Module (Cat Nos. 19932, 19943) different from the earlier Fast Blue Substrate products?
The earlier Fast Blue Substrate products (Cat Nos. QVT0506 and QVT0507) provided only the Blue Buffer and Blue Reagents 1, 2, and 3. The ViewRNA Tissue Assay Blue Module (Cat Nos. 19932, 19943) includes these same components plus a Label Probe 6-AP and a stop reagent.
For Research Use Only. Not for use in diagnostic procedures.