missing translation for 'onlineSavingsMsg'
Learn More

FANCD2 Antibody, HRP, R&D Systems™

Numéro de catalogue. NB100182H
Modifier l'affichage
Click to view available options
Quantité:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
NB100182H 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. NB100182H Fournisseur Bio-Techne Code fournisseur NB100182H

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

FANCD2 Polyclonal specifically detects FANCD2 in Avian, Canine, Human, Kangaroo, Mouse, Primate, Rat, Zebrafish samples. It is validated for ChIP Assay, Flow Cytometry, Gene Knock-Out, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, RNA Inhibition, Western Blot.

Spécifications

Antigène FANCD2
Applications ChIP Assay, Flow Cytometry, Gene Knock-Out, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Western Blot
Classification Polyclonal
Conjugué HRP
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting, Knockout Validated, Knockdown Validated
Formule PBS with No Preservative
Numéro d’acquisition du gène Q9BXW9
Alias du gène DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3
Symboles de gène FANCD2
Espèces hôtes Rabbit
Immunogène This FANCD2 Antibody [HRP] was developed against human FANCD2 fusion protein (N-terminal fragment). [Swiss-Prot #Q9BXW9]
Méthode de purification Affinity Purified
Quantité 0.05 mL
Statut réglementaire RUO
Disciplines de recherche Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents
Primaire ou secondaire Primary
ID du gène (Entrez) 2177
Spécifications de test This antibody is specific for FANCD2.
Espèces cibles Avian, Canine, Rat, Human, Kangaroo, Mouse, Primate, Rat, Zebrafish
Contenu et stockage Store at 4C in the dark.
Type de produit Antibody
Isotype IgG
Gamme de produit Novus
Afficher plus de résultats Afficher moins de résultats

For Research Use Only