missing translation for 'onlineSavingsMsg'
Learn More

Medisca Famotidine USP

Catalog No. 14185659
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185659 25 g
14185251 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14185659 Supplier Medisca Supplier No. 065504

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Famotidine USP is a white to pale yellowish-white, crystalline powder.

Famotidine USP is a white to pale yellowish-white, crystalline powder.

Specifications

Chemical Name or Material Famotidine USP
Packaging Jar 300ml, White, HDPE, Round, 53-400, TRIVENI-DB, CT
CAS 76824-35-6
Type Drug Standard
Molecular Weight (g/mol) 337.45
Quantity 25 g
Percent Purity 99.5 to 101.0 %
Grade USP