missing translation for 'onlineSavingsMsg'
Learn More

Famotidine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015475
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015475 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015475 Supplier Merck Emd Millipore Supplier No. PHR10551G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Famotidine is a histamine H2-receptor antagonist, which promotes the healing of erosive esophagitis, gastric and duodenal ulcers since it inhibits the gastric acid secretion in humans.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 76824-35-6
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C8H15N7O2S3
MDL Number MFCD00079297
Synonym N′-(Aminosulfonyl)-3-([2-(diaminomethyleneamino)-4-thiazolyl]methylthio)propanamidine
Type Certified Reference Material
Quantity 1 g
Formula Weight 337.45
Grade Certified Reference Material