missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAM78A Polyclonal Antibody
GREENER_CHOICE

Numéro de catalogue. PIPA5112974
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PIPA5112974 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PIPA5112974 Fournisseur Invitrogen™ Code fournisseur PA5112974
Il en reste null

Rabbit Polyclonal Antibody

FAM78A Polyclonal Antibody for Western Blot, IHC (P), ELISA

FAM78A is a protein coding gene.

Spécifications

Antigen FAM78A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene FAM78A
Gene Accession No. Q5JUQ0
Gene Alias A130092J06Rik; C9orf59; chromosome 9 open reading frame 59; FAM78A; family with sequence similarity 78 member A; family with sequence similarity 78, member A; protein FAM78A; RGD1566351
Gene Symbols FAM78A
Host Species Rabbit
Immunogen Recombinant Human Protein FAM78A protein (21-283AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286336
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats