missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Falcon™ Sterile Sample Containers
Description
- Sterile, disposable polypropylene containers provide secure sample containment
- Available in 4 1/2 oz. (110 mL) and 8 oz. (220 mL) sizes both with and without matching lids
- Feature molded-in graduations for easy measurements
- Graduated in ounces and milliliters to provide convenient, single-use, timesaving containers for collection, transportation, and storage of a wide variety of specimens
- Inert and chemically resistant to commonly used laboratory reagents at room temperature
Features
Sterile- 4 1/4 oz. graduations from 1/2 oz. to 4 1/2 oz. in 1/4 oz. increments and from 20 mL to 110 mL in 10 mL increments
- 8 oz. size graduated from 1/4 oz. to 8 oz. in 1/4 oz. increments and from 20 mL to 220 mL in 10 mL increments
Spécifications
Spécifications
| Closure Included | No |
| Graduated | Yes |
| Material | Polypropylene |
| Sterility | Sterile |
| Capacity (Metric) | 220 mL |
| No. per Case | 500/Cs. |
| No. per Pack | 20/Pk. |