missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ 384-Well, Cell Culture-Treated, Flat-Bottom Microplate

Catalog No. 08772129
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
08772129 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08772129 Supplier Falcon™ Supplier No. 353988
Only null left

Color:White; Sterility:Sterile; Lid:With Lid

  • Ideal for use in high throughput systems
  • Falcon 384-well microplates are compatible with automation systems and meet standard ANSI/SBS footprint dimensions for microplates
  • Specifications

    Format Standard
    Lid With Lid
    Content And Storage Ambient
    Culture Area 10 mm2
    Array 16 x 24
    Binding Type TC-Treated
    Color White
    For Use With (Equipment) Automation systems
    Material Polystyrene
    No. of Wells 384
    Volume (Metric) Working 15 to 110 μL
    Sterility Sterile
    Surface Treatment Cell Culture
    Well Shape Flat Bottom (F-well)
    No. per Case 50/Cs.
    No. per Pack 5/Pk.
    Quantity 50/Cs.
    Well Color White
    Show More Show Less