Learn More
Invitrogen™ F(ab)-Rabbit anti-Goat IgG Fc Secondary Antibody, Alexa Fluor™ 647
Description
This F(ab)-Rabbit anti-Goat IgG Fc specific antibody product is a rabbit polyclonal antibody F(ab) fragment that is specific to the Fc region of goat IgG. The product has minimal to no cross-reactivity towards polyclonal mouse, rat, guinea pig, bovine, hamster, horse, sheep, human, monkey, donkey IgG, and chicken IgY as tested by direct ELISA. Cross-reactivity towards other immunoglobulins has not been tested. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. F(ab) antibody fragment offer several benefits over whole IgGs, due to their smaller size (~50 kD vs. ~150 kD) and the absence of the Fc region. The smaller size is expected to allow better penetration into tissue sections and improved staining in applications such as IHC. The lack of Fc region results in reduced nonspecific background staining when staining cells/ tissues with Fc receptors (such as immune cells) and other Fc interaction-related interference. Fc specific F(ab) fragments provide a range of multiplexing options beyond what is possible with whole IgGs for applications such as immunocytochemistry, immunohistochemistry, flow cytometry, and western blotting. Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining.
Specifications
Specifications
| Antigen | Goat IgG Fc |
| Applications | Peptide Array, Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 647 |
| Formulation | PBS with 5mM sodium azide; pH 7.5 |
| Host Species | Rabbit |
| Immunogen | Goat IgG Fc. |
| Purification Method | Size-exclusion chromatography |
| Quantity | 50 μg |
| Show More |