missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ F(ab')2-Rabbit anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, AP

Numéro de catalogue. PA184469
Modifier l'affichage
Click to view available options
Quantité:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
PA184469 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PA184469 Fournisseur Invitrogen™ Code fournisseur PA184469
  • ONLINE EXCLUSIVE PRICE
    $962.39
    $559.50 / chaque

    Économisez $402.89

    42.0 % de rabais Les écarts et divulgations suivants peuvent entraîner des prix inférieurs aux rabais indiqués, sans toutefois entraîner de réductions de prix supplémentaires : Le prix affiché reflète une réduction promotionnelle temporaire. Aucun achat additionnel ni engagement commercial n’est requis pour bénéficier du prix réduit. Le prix peut être modifié en tout temps. Le prix promotionnel est valide uniquement pour les commandes en ligne. Votre tarification contractuelle peut différer. D’autres exclusions peuvent s’appliquer.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

F(ab')2-Rabbit Polyclonal Secondary Antibody

Cross reactivity is observed with rat IgG. Cross-reactivity with human IgG has been minimized by solid phase absorption. F(ab')2 fragments were prepared by pepsin digestion of the IgG followed by a gel filtration step to remove the remaining intact IgG or Fc fragments. Rabbit anti Mouse IgG antibody recognizes mouse IgG.Cross reactivity is observed with rat IgG. Cross-reactivity with human IgG has been minimised by solid phase absorption.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Spécifications

Antigène Mouse IgG (H+L) Cross-Adsorbed
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugué Alkaline Phosphatase (AP)
Formule TBS with 25% glycerol, 1% BSA and <0.1% sodium azide; pH 8.0
Espèces hôtes Rabbit
Immunogène Murine IgG.
Méthode de purification Affinity chromatography
Quantité 500 μg
Statut réglementaire RUO
Primaire ou secondaire Secondary
Espèces cibles Mouse
Contenu et stockage 4°C, do not freeze
Type de produit Secondary Antibody
Forme Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats