missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EZH1 Polyclonal Antibody

Description
This antibody is 86% homologous to rat and canine. Suggested positive control: antigen standard for EZH1 (transient overexpression lysate), human or mouse spleen.
EZH1 is a component of a noncanonical Polycomb repressive complex-2 (PRC2) that mediates methylation of histone H3 lys27 (H3K27) and functions in the maintenance of embryonic stem cell pluripotency and plasticity (Shen et al., 2008 ).
Spécifications
Spécifications
| Antigène | EZH1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 0.05% sodium azide |
| Gène | EZH1 |
| Numéro d’acquisition du gène | P70351, Q92800 |
| Alias du gène | enhancer of zeste 1 polycomb repressive complex 2 subunit; enhancer of zeste homolog 1; enhancer of zeste homolog 1 (Drosophila); Enx2; ENX-2; Ezh1; Histone-lysine N-methyltransferase EZH1; hypothetical protein LOC664754; KIAA0388; KMT6B; si:ch211-250i3.1; zgc:136829 |
| Symboles de gène | EZH1 |
| Afficher plus de résultats |