Learn More
Foxx Life Sciences EZB Pr S 5L PP NS NoCap 12PK
GREENER_CHOICE
Numéro de catalogue. 502236444
Encompass
EZBio pure S Bottles (PP/HDPE) offer durable chemical-resistant storage for reagents buffers and media in laboratory and bioprocessing environments Featuring the GL45 VersaCap system for a leak-resistant seal these bottles include molded graduation marks for precise volume reference and seamless compatibility with patented VersaCap closures br br Features Br Chemical-resistant PP or HDPE material for safe storage Br Square profile design for space-efficient storage and improved shelf utilization br GL45 VersaCap with molded rotating adapter to reduce tubing tangling br Made entirely of 100% USP Class VI Components br Clear molded graduation marks for easy volume measurement and reference br Manufactured in a Class 7 cleanroom at an ISO 13485 certified facility br br Applications br Reagent storage br Media and buffer preparation br Sample handling and transfer br Feed and intermediate solution handling br Single-use fluid management workflows
Numéro de catalogue. 502236444 Fournisseur Foxx Life Sciences Code fournisseur 5052F31FLS
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
EZBio pure S Bottles (PP/HDPE) offer durable chemical-resistant storage for reagents buffers and media in laboratory and bioprocessing environments Featuring the GL45 VersaCap system for a leak-resistant seal these bottles include molded graduation marks for precise volume reference and seamless compatibility with patented VersaCap closures br br Features Br Chemical-resistant PP or HDPE material for safe storage Br Square profile design for space-efficient storage and improved shelf utilization br GL45 VersaCap with molded rotating adapter to reduce tubing tangling br Made entirely of 100% USP Class VI Components br Clear molded graduation marks for easy volume measurement and reference br Manufactured in a Class 7 cleanroom at an ISO 13485 certified facility br br Applications br Reagent storage br Media and buffer preparation br Sample handling and transfer br Feed and intermediate solution handling br Single-use fluid management workflows