missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EXPRESS SYBR™ GreenER™ qPCR Supermix, with premixed ROX
Kits are compatible with fast-cycling instruments for SYBR-based qPCR experiments.
Specifications
| Detection Method | SYBR |
|---|---|
| GC-Rich PCR Performance | High |
| PCR Method | qPCR |
| Polymerase | Taq DNA Polymerase |
| Reaction Speed | Fast or Standard |
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
11794200
|
Invitrogen™
11794200 |
200 Reactions |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
1179401K
|
Invitrogen™
1179401K |
1000 Reactions |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This EXPRESS SYBR GreenER qPCR SuperMix comes with premixed ROX reference dye.
- Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
- SYBR GreenER dye
- UDG carryover protection - uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination
Specifications
| SYBR | |
| qPCR | |
| Fast or Standard | |
| SYBR GreenER | |
| Gene Expression |
| High | |
| Taq DNA Polymerase | |
| 7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf™ MasterCycler™ RealPlex, StepOne™, Fast Mode, StepOne™, Standard Mode | |
| Real Time PCR SYBR Master Mix | |
| DNA (Genomic), cDNA |
For Research Use Only. Not for use in diagnostic procedures.