Learn More
Invitrogen™ EXPRESS SYBR™ GreenER™ qPCR Supermix, with premixed ROX
Kits are compatible with fast-cycling instruments for SYBR-based qPCR experiments.
Supplier:  Invitrogen™ 1179401K
Encompass
 View more versions of this product
Catalog No. 1179401K
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
This EXPRESS SYBR GreenER qPCR SuperMix comes with premixed ROX reference dye.
  • Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
  • SYBR GreenER dye
  • UDG carryover protection - uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination
Specifications
Chemical Identifiers
SYBR
qPCR
Fast or Standard
SYBR GreenER
Gene Expression
High
Taq DNA Polymerase
7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf™ MasterCycler™ RealPlex, StepOne™, Fast Mode, StepOne™, Standard Mode
Real Time PCR SYBR Master Mix
DNA (Genomic), cDNA
Specifications
Contains 5 × 5 ml EXPRESS SYBR™ GreenER™ qPCR SuperMix with Premixed ROX. Store at −20°C for long-term storage. EXPRESS qPCR SuperMixes may be stored at 4–8°C for up to one month.
High
Taq DNA Polymerase
7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf™ MasterCycler™ RealPlex, StepOne™, Fast Mode, StepOne™, Standard Mode
SYBR GreenER
1,000 reactions
Gene Expression
DNA (Genomic), cDNA
SYBR
qPCR
Fast or Standard
Fast-Cycling Instrument Kit
Real Time PCR SYBR Master Mix
1000 Reactions
1000 Reactions
5 x 5 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.