missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EXPRESS One-Step SYBR™ GreenER™ Kit, with premixed ROX
$2,370.00 - $10,210.00
Specifications
| Content And Storage | EXPRESS One-Step SYBR™ GreenER™ SuperMix Kits are composed of 2 modules: EXPRESS SYBR™ GreenER™ RT module (containing SuperScript™ III Reverse Transcriptase and RNAseOUT™) and EXPRESS SYBR™ GreenER™ qPCR SuperMix with ROX premixed. Store at -20°C upon receipt. Once thawed, the EXPRESS SYBR™ GreenER™ qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteed to be stable for 6 months when properly stored. |
|---|---|
| Detection Method | SYBR |
| Format | Tube |
| PCR Method | 1-step RT-qPCR |
| Polymerase | Taq DNA Polymerase |
| missing translation for 'productCode' | missing translation for 'brand' | No. of Reactions | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| missing translation for 'productCode' | missing translation for 'brand' | No. of Reactions | Price | Quantity & Availability | |||||
11790200
|
Invitrogen™
11790200 |
500 Reactions |
Each for $2,370.00
|
|
|||||
1179001K
|
Invitrogen™
1179001K |
2500 Reactions |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This kit comes with premixed ROX reference dye. EXPRESS One-Step SYBR GreenER SuperMix kits with a separate tube of ROX reference dye are also available.
- Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
- The One-Step SuperScript™ III Reverse Transcriptase for one-step qRT-PCR
- SYBR GreenER dye
- UDG carryover protection - uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination
Specifications
| EXPRESS One-Step SYBR™ GreenER™ SuperMix Kits are composed of 2 modules: EXPRESS SYBR™ GreenER™ RT module (containing SuperScript™ III Reverse Transcriptase and RNAseOUT™) and EXPRESS SYBR™ GreenER™ qPCR SuperMix with ROX premixed. Store at -20°C upon receipt. Once thawed, the EXPRESS SYBR™ GreenER™ qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteed to be stable for 6 months when properly stored. |
|
| Tube | |
| Taq DNA Polymerase | |
| 7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf™ MasterCycler™ RealPlex, StepOne™, Fast Mode, StepOne™, Standard Mode | |
| Gene Expression |
| SYBR | |
| 1-step RT-qPCR | |
| Fast or Standard | |
| SYBR GreenER | |
| RNA |
For Research Use Only. Not for use in diagnostic procedures.