Learn More
Invitrogen™ EXPRESS One-Step SYBR™ GreenER™ Kit, with premixed ROX
Supplier:  Invitrogen™ 1179001K
Encompass
 View more versions of this product
Catalog No. 1179001K
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
This kit comes with premixed ROX reference dye. EXPRESS One-Step SYBR GreenER SuperMix kits with a separate tube of ROX reference dye are also available.
  • Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
  • The One-Step SuperScript™ III Reverse Transcriptase for one-step qRT-PCR
  • SYBR GreenER dye
  • UDG carryover protection - uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination
Specifications
Chemical Identifiers
EXPRESS One-Step SYBR™ GreenER™ SuperMix Kits are composed of 2 modules: EXPRESS SYBR™ GreenER™ RT module (containing SuperScript™ III Reverse Transcriptase and RNAseOUT™) and EXPRESS SYBR™ GreenER™ qPCR SuperMix with ROX premixed.
Store at -20°C upon receipt. Once thawed, the EXPRESS SYBR™ GreenER™ qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteed to be stable for 6 months when properly stored.
Tube
Taq DNA Polymerase
7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf™ MasterCycler™ RealPlex, StepOne™, Fast Mode, StepOne™, Standard Mode
Gene Expression
SYBR
1-step RT-qPCR
Fast or Standard
SYBR GreenER
RNA
Specifications
EXPRESS One-Step SYBR™ GreenER™ SuperMix Kits are composed of 2 modules: EXPRESS SYBR™ GreenER™ RT module (containing SuperScript™ III Reverse Transcriptase and RNAseOUT™) and EXPRESS SYBR™ GreenER™ qPCR SuperMix with ROX premixed.
Store at -20°C upon receipt. Once thawed, the EXPRESS SYBR™ GreenER™ qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteed to be stable for 6 months when properly stored.
Tube
Taq DNA Polymerase
7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf™ MasterCycler™ RealPlex, StepOne™, Fast Mode, StepOne™, Standard Mode
SYBR GreenER
2,500 reactions
Gene Expression
RNA
SYBR
1-step RT-qPCR
Fast or Standard
EXPRESS One-Step SYBR GreenER Kit
Real Time PCR SYBR Master Mix
2500 Reactions
2500 Reactions
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.