missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Expi293™ PRO Transfection Kit

Numéro de catalogue. a40002930
Change view
Click to view available options
Sufficient For:
For 1 L transfection
For 10 L transfection
For 50 L transfection
For 6 L transfection
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Sufficient For
A40002930 For 50 L transfection
A40002926 For 1 L transfection
A40002929 For 6 L transfection
A40002927 For 10 L transfection
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. A40002930 Fournisseur Gibco™ Code fournisseur A40002930

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

The Expi293 PRO Transfection Kit enables high-efficiency delivery of DNA into HEK293 cells, supporting fast, scalable protein expression. Optimized for use with Expi293F cells, it delivers robust performance across a broad range of protein types.

The Expi293 PRO Transfection Kit is a core component of the Expi293 PRO Expression System. It is designed for transient transfection of high density cultures of HEK 293 cells. The high-efficiency, cationic, lipid-based transfection reagent and transfection enhancers are designed to power the highest possible level of protein expression from Expi293 PRO cells cultured in Expi293 PRO Expression Medium.

  • Designed specifically for transfection of high-density suspension cell culture, with complimentary feed and enhancers that boost transfection performance and protein expression
  • Achieves protein yields 2- to 10-fold higher than other transfection reagents used on high-density Expi293 PRO cell cultures
  • Provides robust and reproducible transfection results, giving users greater confidence in their results
  • Scale transfections for culture volumes of less than 1 mL to greater than 10 liters, while maintaining equivalent volumetric protein yields

The Expi293 PRO Transfection Kit is specifically designed to work with Expi293 PRO cells in the Expi29 PRO Expression System. The transfection kit consists of the cationic lipid-based Expi293 PRO transfection reagent and Expi293 PRO enhancer and feed, which boost transfection, cell viability, and protein expression.

Spécifications

Cell Type HEK293
Format Kit
Sample Type Plasmid DNA
Promoter pcDNA™ 3.4
Transfection Technique Lipid-based Transfection, Transient Transfection
Product Line Expi293 PRO
Product Type Transfection Kit
Purity Cell Culture Grade
Quantity For 50 L transfection
Sufficient For For 50 L transfection

For Research Use Only. Not for use in diagnostic procedures.